siRNA Details
| Gene Symbol: PCDHB7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018940.3 | NP_061763.1 | 1 | 460 | ||
| GenBank mRNA | AF152500.1 | - | 1 | 245 | ||
| AF217750.1 | - | 1 | 245 | |||
| AK024641.1 | - | 1 | 419 | |||
| AK222560.1 | - | 1 | 421 | |||
| AK222678.1 | - | 1 | 405 | |||
| AK315054.1 | - | 1 | 463 | |||
| BC098574.1 | - | 1 | 399 | 30925247 | ||
| BC130331.1 | - | 1 | 327 | 40146362 | ||
| BC130333.1 | - | 1 | 327 | 40146364 | ||
| DA779188.1 | - | 1 | 461 | |||

PCDHB7
Genomic Map