siRNA Details
| Gene Symbol: PCDHB6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018939.3 | NP_061762.2 | 1 | 312 | ||
| GenBank mRNA | AF152499.1 | - | 1 | 76 | ||
| AF217752.1 | - | 1 | 76 | |||
| AK313482.1 | - | 1 | 233 | |||
| DA789782.1 | - | 1 | 312 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PCDHB6| Gene Symbol: PCDHB6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018939.3 | NP_061762.2 | 1 | 312 | ||
| GenBank mRNA | AF152499.1 | - | 1 | 76 | ||
| AF217752.1 | - | 1 | 76 | |||
| AK313482.1 | - | 1 | 233 | |||
| DA789782.1 | - | 1 | 312 | |||