siRNA Details
| Gene Symbol: PCDHB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018936.3 | NP_061759.1 | 1 | 1046 | ||
| GenBank mRNA | AB209745.1 | - | 1 | 822 | ||
| AF152495.1 | - | 1 | 862 | |||
| AF217756.1 | - | 1 | 862 | |||
| AK027526.1 | - | 1 | 1017 | |||
| AK314642.1 | - | 1 | 1000 | |||
| BC098347.1 | - | 1 | 909 | 40007951 | ||
| BC098575.1 | - | 1 | 1010 | 30921306 | ||

PCDHB2
Genomic Map