siRNA Details
| Gene Symbol: TEX15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031271.3 | NP_112561.2 | 1 | 4379 | ||
| XM_005273575.3 | XP_005273632.1 | 8 | 5747 | |||
| XM_005273576.3 | XP_005273633.1 | 5 | 5309 | |||
| XM_006716369.3 | XP_006716432.1 | 7 | 5639 | |||
| XM_011544588.2 | XP_011542890.1 | 7 | 5603 | |||
| XM_011544589.2 | XP_011542891.1 | 8 | 5712 | |||
| XM_011544590.2 | XP_011542892.1 | 8 | 5978 | |||
| XM_011544591.2 | XP_011542893.1 | 7 | 5640 | |||
| XM_011544592.2 | XP_011542894.1 | 7 | 5578 | |||
| XM_011544593.2 | XP_011542895.1 | 5 | 6535 | |||
| GenBank mRNA | AF285605.1 | - | 1 | 4379 | ||
| BC140934.1 | - | 1 | 4454 | 9056746 | ||

TEX15
Genomic Map