siRNA Details
| Gene Symbol: CRTAM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304782.1 | NP_001291711.1 | 5 | 663 | ||
| NM_019604.3 | NP_062550.2 | 10 | 1211 | |||
| XM_011542900.2 | XP_011541202.1 | 9 | 1137 | |||
| GenBank mRNA | AB209830.1 | - | 5 | 572 | ||
| AF001622.1 | - | 10 | 1162 | |||
| AK293470.1 | - | 5 | 592 | |||
| AK313578.1 | - | 10 | 1209 | |||
| BC070266.1 | - | 10 | 1211 | 30418882 | ||
| BX118379.1 | - | 4 | 378 | |||

CRTAM
Genomic Map