siRNA Details
| Gene Symbol: SERTAD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019605.3 | NP_062551.1 | 4 | 567 | ||
| GenBank mRNA | AK096381.1 | - | 4 | 533 | ||
| AK315386.1 | - | 3 | 566 | |||
| BC012083.1 | - | 4 | 567 | 4564813 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SERTAD4| Gene Symbol: SERTAD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019605.3 | NP_062551.1 | 4 | 567 | ||
| GenBank mRNA | AK096381.1 | - | 4 | 533 | ||
| AK315386.1 | - | 3 | 566 | |||
| BC012083.1 | - | 4 | 567 | 4564813 | ||