siRNA Details
| Gene Symbol: CPXM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001184699.1 | NP_001171628.1 | 14 | 1945 | ||
| NM_019609.4 | NP_062555.1 | 14 | 2167 | |||
| GenBank mRNA | AI470029.1 | - | 1 | 239 | ||
| AK027661.1 | - | 14 | 2103 | |||
| AK075508.1 | - | 10 | 1379 | |||
| AK075527.1 | - | 14 | 1876 | |||
| AK303142.1 | - | 11 | 1646 | |||
| AK315027.1 | - | 14 | 1932 | |||
| AW874578.1 | - | 1 | 236 | |||
| AY358956.1 | - | 14 | 2123 | |||
| BC063430.1 | - | 14 | 2144 | 5239864 | ||

CPXM1
Genomic Map