siRNA Details
| Gene Symbol: WDR45B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019613.3 | NP_062559.2 | 4 | 460 | ||
| XM_005256377.4 | XP_005256434.1 | 3 | 283 | |||
| XM_017024862.1 | XP_016880351.1 | 4 | 375 | |||
| XM_017024863.1 | XP_016880352.1 | 4 | 219 | |||
| GenBank mRNA | AF091083.1 | - | 2 | 103 | ||
| AK297477.1 | - | 4 | 214 | |||
| AM182326.1 | - | 4 | 265 | |||
| AY691427.1 | - | 2 | 91 | |||
| BC000974.2 | - | 4 | 243 | |||
| BC007838.1 | - | 4 | 432 | |||
| CR456770.1 | - | 2 | 91 | |||

WDR45B
Genomic Map