siRNA Details
| Gene Symbol: TRPV5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019841.6 | NP_062815.3 | 7 | 1153 | ||
| GenBank mRNA | AF304464.1 | - | 7 | 1238 | ||
| AJ271207.2 | - | 7 | 889 | |||
| AJ487965.2 | - | 7 | 891 | |||
| BC034740.1 | - | 7 | 1153 | 5186668 | ||

TRPV5
Genomic Map