siRNA Details
| Gene Symbol: ERO1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019891.3 | NP_063944.3 | 13 | 1331 | ||
| GenBank mRNA | AF252538.1 | - | 13 | 1248 | ||
| AK315351.1 | - | 13 | 1250 | |||
| BC032823.2 | - | 13 | 1182 | 4829502 | ||
| BC044573.1 | - | 13 | 1277 | 5270105 | ||

ERO1B
Genomic Map