siRNA Details
| Gene Symbol: SLC2A9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001290.1 | NP_001001290.1 | 13 | 1667 | ||
| NM_020041.2 | NP_064425.2 | 12 | 1656 | |||
| XM_017008460.1 | XP_016863949.1 | 11 | 3313 | |||
| GenBank mRNA | AF210317.1 | - | 12 | 1649 | ||
| BC018897.2 | - | 13 | 1667 | 3949549 | ||
| BC110414.1 | - | 12 | 1686 | 5762360 | ||

SLC2A9
Genomic Map