siRNA Details
| Gene Symbol: LGALS14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020129.2 | NP_064514.1 | 3 | 401 | ||
| NM_203471.1 | NP_982297.1 | 4 | 706 | |||
| GenBank mRNA | AF123762.1 | - | 2 | 178 | ||
| AF267852.1 | - | 3 | 401 | |||
| AF367430.1 | - | 3 | 401 | |||
| AF367431.1 | - | 3 | 401 | |||
| AF367432.1 | - | 3 | 401 | |||
| AI167958.1 | - | 2 | 373 | |||
| AK312040.1 | - | 3 | 240 | |||
| BC022257.1 | - | 3 | 257 | 4770140 | ||
| BG434228.1 | - | 4 | 532 | |||
| BQ007347.1 | - | 2 | 390 | |||
| FJ613341.1 | - | 3 | 205 | |||
| FJ613342.1 | - | 3 | 205 | |||

LGALS14
Genomic Map