siRNA Details
| Gene Symbol: STARD7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020151.3 | NP_064536.2 | 4, 5 | 1069 | ||
| XM_011511499.1 | XP_011509801.1 | 4, 5 | 686 | |||
| GenBank mRNA | AF270647.1 | - | 4, 5 | 996 | ||
| AK054615.1 | - | 4, 5 | 435 | |||
| AK291190.1 | - | 4, 5 | 451 | |||
| BC003028.1 | - | 4, 5 | 1052 | |||
| BC007894.2 | - | 5, 6 | 827 | 4134933 | ||
| BC008894.2 | - | 4, 5 | 1052 | 3833465 | ||
| BC009998.2 | - | 5, 6 | 827 | 4128869 | ||
| BC012774.2 | - | 4, 5 | 1019 | 3842611 | ||
| BC012793.2 | - | 4, 5 | 1033 | 4080200 | ||
| BC013279.2 | - | 4, 5 | 1052 | 3830320 | ||
| BC014076.2 | - | 4, 5 | 1029 | 4127007 | ||
| BC014274.2 | - | 4, 5 | 1048 | 3677198 | ||
| BC032106.1 | - | 4, 5 | 1043 | 4643387 | ||

STARD7
Genomic Map