siRNA Details
| Gene Symbol: CCDC177 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271507.1 | NP_001258436.1 | 2 | 3083 | ||
| GenBank mRNA | AF284217.1 | - | 1 | 292 | ||
| AK312175.1 | - | 1 | 406 | |||
| BC105025.1 | - | 1 | 215 | |||
| BC105027.1 | - | 1 | 215 | |||

CCDC177
Genomic Map