siRNA Details
| Gene Symbol: C5orf15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020199.2 | NP_064584.1 | 2 | 462 | ||
| GenBank mRNA | AF226055.1 | - | 2 | 535 | ||
| AK074963.1 | - | 2 | 374 | |||
| AK315362.1 | - | 2 | 360 | |||
| AY157581.1 | - | 2 | 371 | |||
| BC020875.1 | - | 2 | 353 | 4610607 | ||
| BP245947.1 | - | 2 | 462 | |||

C5orf15
Genomic Map