siRNA Details
| Gene Symbol: OTUD7B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020205.3 | NP_064590.2 | 6 | 992 | ||
| XM_011509782.1 | XP_011508084.1 | 6 | 989 | |||
| XM_011509783.2 | XP_011508085.1 | 6 | 1219 | |||
| XM_011509784.2 | XP_011508086.1 | 6 | 783 | |||
| XM_011509785.2 | XP_011508087.1 | 6 | 4416 | |||
| XM_011509786.1 | XP_011508088.1 | 6 | 855 | |||
| XM_011509787.1 | XP_011508089.1 | 6 | 598 | |||
| XM_011509788.1 | XP_011508090.1 | 5 | 707 | |||
| XM_017001850.1 | XP_016857339.1 | 6 | 855 | |||
| XM_017001851.1 | XP_016857340.1 | 5 | 841 | |||
| GenBank mRNA | AJ293573.1 | - | 6 | 835 | ||
| AK299790.1 | - | 6 | 978 | |||
| BC020622.1 | - | 6 | 778 | |||
| BC072681.1 | - | 6 | 929 | 6170513 | ||
| BX641079.1 | - | 4 | 427 | |||

OTUD7B
Genomic Map