siRNA Details
| Gene Symbol: ERCC6L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020207.4 | NP_064592.2 | 19 | 4883 | ||
| XM_011518641.2 | XP_011516943.1 | 18 | 4730 | |||
| XM_011518644.2 | XP_011516946.1 | 18 | 4675 | |||
| XM_011518648.2 | XP_011516950.1 | 10 | 2955 | |||
| XM_011518650.2 | XP_011516952.1 | 19 | 4202 | |||
| XM_017014707.1 | XP_016870196.1 | 19 | 4870 | |||
| GenBank mRNA | AK095025.1 | - | 1 | 693 | ||
| BC022957.1 | - | 4 | 1312 | 4995030 | ||

ERCC6L2
Genomic Map