siRNA Details
| Gene Symbol: ERCC6L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020207.4 | NP_064592.2 | 18 | 4052 | ||
| XM_011518641.2 | XP_011516943.1 | 17 | 3899 | |||
| XM_011518644.2 | XP_011516946.1 | 17 | 3844 | |||
| XM_011518645.2 | XP_011516947.1 | 18 | 4052 | |||
| XM_011518648.2 | XP_011516950.1 | 9 | 2124 | |||
| XM_011518650.2 | XP_011516952.1 | 18 | 3371 | |||
| XM_017014707.1 | XP_016870196.1 | 18 | 4039 | |||
| GenBank mRNA | AK308131.1 | - | 3 | 1087 | ||
| AL133663.1 | - | 13 | 2652 | |||
| BC022957.1 | - | 3 | 481 | 4995030 | ||
| BG108141.1 | - | 2 | 312 | |||
| BQ013632.1 | - | 1 | 228 | |||
| CA425791.1 | - | 1 | 220 | |||

ERCC6L2
Genomic Map