siRNA Details
| Gene Symbol: WDR93 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001284395.1 | NP_001271324.1 | 13, 14 | 1567 | ||
| NM_020212.1 | NP_064597.1 | 13, 14 | 1651 | |||
| GenBank mRNA | AK097525.1 | - | 13, 14 | 1535 | ||
| AK302032.1 | - | 7, 8 | 1056 | |||
| AL390084.1 | - | 5, 6 | 635 | |||
| AL390085.1 | - | 5, 6 | 635 | |||
| BC064626.1 | - | 13, 14 | 1651 | 5742386 | ||

WDR93
Genomic Map