siRNA Details
| Gene Symbol: SDHAF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020186.2 | NP_064571.1 | 1 | 1155 | ||
| GenBank mRNA | AF201933.1 | - | 1 | 44 | ||
| BC020621.1 | - | 1 | 37 | 4766494 | ||
| BC022858.1 | - | 1 | 45 | 4132451 | ||
| BC028409.1 | - | 1 | 1163 | |||

SDHAF3
Genomic Map