siRNA Details
| Gene Symbol: YAE1D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282446.1 | NP_001269375.1 | 1, 2 | 167 | ||
| NM_020192.4 | NP_064577.1 | 1, 2 | 167 | |||
| GenBank mRNA | AF226046.1 | - | 1, 2 | 133 | ||
| AK293511.1 | - | 1, 2 | 137 | |||
| AK314937.1 | - | 1, 2 | 139 | |||
| BC022043.1 | - | 1, 2 | 137 | 4734433 | ||
| BM767075.1 | - | 1, 2 | 167 | |||
| CR457198.1 | - | 1, 2 | 124 | |||

YAE1D1
Genomic Map