siRNA Details
| Gene Symbol: YAE1D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020192.4 | NP_064577.1 | 3 | 429 | ||
| GenBank mRNA | AF226046.1 | - | 3 | 395 | ||
| AK314937.1 | - | 3 | 401 | |||
| BC022043.1 | - | 3 | 399 | 4734433 | ||
| BM767075.1 | - | 3 | 429 | |||
| BX648555.1 | - | 1 | 2982 | |||
| CR457198.1 | - | 3 | 386 | |||

YAE1D1
Genomic Map