siRNA Details
| Gene Symbol: COQ9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020312.3 | NP_064708.1 | 2 | 219 | ||
| GenBank mRNA | AF161444.1 | - | 2 | 150 | ||
| AK075438.1 | - | 2 | 220 | |||
| AK290627.1 | - | 2 | 149 | |||
| AK293585.1 | - | 2 | 153 | |||
| AK295793.1 | - | 2 | 162 | |||
| AK303527.1 | - | 2 | 142 | |||
| BC029341.1 | - | 2 | 153 | |||
| BC054340.1 | - | 2 | 153 | 6514554 | ||
| BC064946.1 | - | 2 | 154 | 6023461 | ||
| BM712215.1 | - | 2 | 207 | |||

COQ9
Genomic Map