siRNA Details
| Gene Symbol: PITHD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020362.4 | NP_065095.2 | 3 | 407 | ||
| XM_011541804.1 | XP_011540106.1 | 2 | 332 | |||
| GenBank mRNA | AF218024.1 | - | 3 | 388 | ||
| AF221595.1 | - | 3 | 399 | |||
| AF271784.1 | - | 3 | 403 | |||
| AK296662.1 | - | 2 | 332 | |||
| AK310330.1 | - | 3 | 354 | |||
| AK313005.1 | - | 3 | 375 | |||
| BC017208.2 | - | 3 | 368 | 3943360 | ||
| DB104988.1 | - | 3 | 407 | |||

PITHD1
Genomic Map