siRNA Details
| Gene Symbol: C12orf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304811.1 | NP_001291740.1 | 10 | 1325 | ||
| NM_020374.3 | NP_065107.1 | 10 | 1333 | |||
| XM_005253716.1 | XP_005253773.1 | 9 | 1204 | |||
| XM_006718992.3 | XP_006719055.1 | 10 | 1333 | |||
| GenBank mRNA | AJ272205.1 | - | 10 | 1276 | ||
| AK024811.1 | - | 2 | 174 | |||
| AK074479.1 | - | 10 | 1333 | |||
| AL834377.1 | - | 10 | 1296 | |||
| BC016908.1 | - | 10 | 1276 | 4402263 | ||
| BX641128.1 | - | 10 | 1325 | |||

C12orf4
Genomic Map