siRNA Details
| Gene Symbol: CYSLTR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308465.1 | NP_001295394.1 | 6 | 1361 | ||
| NM_001308467.1 | NP_001295396.1 | 6 | 1338 | |||
| NM_001308468.1 | NP_001295397.1 | 4 | 1159 | |||
| NM_001308469.1 | NP_001295398.1 | 4 | 1136 | |||
| NM_001308470.1 | NP_001295399.1 | 3 | 1069 | |||
| NM_001308471.1 | NP_001295400.1 | 2 | 968 | |||
| NM_001308476.1 | NP_001295405.1 | 5 | 1232 | |||
| NM_020377.3 | NP_065110.1 | 5 | 1209 | |||
| GenBank mRNA | AB038269.1 | - | 1 | 923 | ||
| AB041947.1 | - | 1 | 994 | |||
| AK291739.1 | - | 5 | 1209 | |||
| AY853711.1 | - | 4 | 888 | |||
| BC069160.1 | - | 1 | 663 | 7216884 | ||
| BC096832.1 | - | 1 | 776 | 6971946 | ||
More Information
Set Membership: |
|

CYSLTR2
Genomic Map