siRNA Details
| Gene Symbol: TRPC7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020389.2 | NP_065122.1 | 3, 4 | 1188 | ||
| GenBank mRNA | AJ272034.1 | - | 2, 3 | 966 | ||
| AJ421783.1 | - | 2, 3 | 966 | |||
| BC128185.1 | - | 2, 3 | 966 | 40113370 | ||
| BC128186.1 | - | 2, 3 | 966 | |||

TRPC7
Genomic Map