siRNA Details
| Gene Symbol: PGLYRP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020393.3 | NP_065126.2 | 6 | 907 | ||
| XM_011509789.2 | XP_011508091.1 | 6 | 919 | |||
| XM_011509790.1 | XP_011508092.1 | 6 | 907 | |||
| XM_011509791.2 | XP_011508093.1 | 6 | 898 | |||
| XM_011509792.1 | XP_011508094.1 | 6 | 907 | |||
| XM_011509793.2 | XP_011508095.1 | 3 | 595 | |||
| GenBank mRNA | AF242518.1 | - | 6 | 623 | ||
| AK292203.1 | - | 6 | 910 | |||
| AY035377.1 | - | 6 | 612 | |||
| BC107157.1 | - | 5 | 594 | 40032748 | ||
| BC107158.1 | - | 5 | 594 | 40032750 | ||
| BC142636.1 | - | 6 | 882 | 40147772 | ||

PGLYRP4
Genomic Map