siRNA Details
| Gene Symbol: CD248 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020404.2 | NP_065137.1 | 1 | 2180 | ||
| GenBank mRNA | AF279142.1 | - | 1 | 2168 | ||
| AJ295846.1 | - | 1 | 2180 | |||
| AK027290.1 | - | 1 | 1302 | |||
| BC051340.1 | - | 1 | 2167 | 6339237 | ||
| BC104484.1 | - | 1 | 2180 | 40007855 | ||
| BC105633.1 | - | 1 | 2180 | 40007854 | ||
| BQ005949.1 | - | 1 | 393 | |||

CD248
Genomic Map