siRNA Details
| Gene Symbol: MAN1C1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289010.1 | NP_001275939.1 | 4, 5 | 1162 | ||
| NM_020379.3 | NP_065112.1 | 4, 5 | 1162 | |||
| XM_017001860.1 | XP_016857349.1 | 3, 4 | 265 | |||
| GenBank mRNA | AB209275.1 | - | 4, 5 | 510 | ||
| AF261655.1 | - | 4, 5 | 1162 | |||
| AK128560.1 | - | 3, 4 | 230 | |||
| BC137017.1 | - | 4, 5 | 968 | 9021010 | ||

MAN1C1
Genomic Map