siRNA Details
| Gene Symbol: LYZL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199951.2 | NP_001186880.1 | 2 | 244 | ||
| NM_020426.3 | NP_065159.1 | 2 | 358 | |||
| GenBank mRNA | AY359018.1 | - | 1 | 162 | ||
| AY742214.1 | - | 1 | 134 | |||
| BC054481.1 | - | 2 | 204 | 6620264 | ||
| BG195727.1 | - | 1 | 212 | |||
| DB462957.1 | - | 2 | 346 | |||
| GQ472214.1 | - | 2 | 204 | |||
| HY024713.1 | - | 2 | 362 | |||

LYZL6
Genomic Map