siRNA Details
| Gene Symbol: DOLPP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135917.1 | NP_001129389.1 | 4 | 343 | ||
| NM_020438.4 | NP_065171.2 | 4 | 343 | |||
| NR_072983.1 | 3 | 258 | ||||
| GenBank mRNA | AK290713.1 | - | 4 | 343 | ||
| AK293627.1 | - | 4 | 334 | |||
| AY189675.1 | - | 4 | 346 | |||
| BC009493.2 | - | 4 | 313 | |||
| BC033686.1 | - | 4 | 320 | 5195127 | ||
| BC040152.1 | - | 3 | 228 | |||
| DA590931.1 | - | 4 | 343 | |||

DOLPP1
Genomic Map