siRNA Details
| Gene Symbol: VN1R1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020633.3 | NP_065684.1 | 1 | 1136 | ||
| GenBank mRNA | AF255342.1 | - | 1 | 883 | ||
| AF302903.1 | - | 1 | 883 | |||
| AK094452.1 | - | 1 | 1542 | |||
| BC095508.1 | - | 1 | 926 | 6971941 | ||

VN1R1
Genomic Map