siRNA Details
| Gene Symbol: VN1R1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020633.3 | NP_065684.1 | 1 | 1232 | ||
| GenBank mRNA | AF255342.1 | - | 1 | 979 | ||
| AF302903.1 | - | 1 | 979 | |||
| AK094452.1 | - | 1 | 1638 | |||
| BC095508.1 | - | 1 | 1022 | 6971941 | ||
| BG167434.1 | - | 2 | 341 | |||

VN1R1
Genomic Map