siRNA Details
| Gene Symbol: MCOLN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020533.2 | NP_065394.1 | 3 | 390 | ||
| GenBank mRNA | AF249319.1 | - | 3 | 333 | ||
| AF287269.1 | - | 3 | 374 | |||
| AJ293659.1 | - | 3 | 329 | |||
| AJ293970.1 | - | 3 | 329 | |||
| AK026102.1 | - | 3 | 344 | |||
| AK222673.1 | - | 3 | 369 | |||
| AK294330.1 | - | 3 | 355 | |||
| BC005149.2 | - | 3 | 372 | 3507836 | ||

MCOLN1
Genomic Map