siRNA Details
| Gene Symbol: ZNF248 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001267597.1 | NP_001254526.1 | 6 | 920 | ||
| NM_021045.2 | NP_066383.1 | 6 | 1292 | |||
| XM_006717485.3 | XP_006717548.1 | 5 | 1249 | |||
| XM_011519597.2 | XP_011517899.1 | 5 | 841 | |||
| XM_017016444.1 | XP_016871933.1 | 4 | 898 | |||
| XM_017016445.1 | XP_016871934.1 | 5 | 1389 | |||
| XM_017016446.1 | XP_016871935.1 | 6 | 940 | |||
| GenBank mRNA | AJ491695.1 | - | 6 | 1214 | ||
| AK299357.1 | - | 6 | 920 | |||
| BC132915.1 | - | 6 | 950 | 40146937 | ||
| BC132917.1 | - | 6 | 950 | 40146939 | ||

ZNF248
Genomic Map