siRNA Details
| Gene Symbol: PPP4R3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122964.2 | NP_001116436.2 | 3 | 712 | ||
| NM_001282850.1 | NP_001269779.1 | 3 | 712 | |||
| NM_020463.3 | NP_065196.1 | 3 | 712 | |||
| XM_005264442.2 | XP_005264499.1 | 3 | 712 | |||
| XM_005264444.2 | XP_005264501.1 | 3 | 712 | |||
| XM_005264445.2 | XP_005264502.1 | 3 | 712 | |||
| XM_005264446.2 | XP_005264503.1 | 3 | 712 | |||
| XM_005264447.2 | XP_005264504.1 | 3 | 712 | |||
| XM_017004531.1 | XP_016860020.1 | 3 | 712 | |||
| XM_017004532.1 | XP_016860021.1 | 3 | 712 | |||
| XM_017004533.1 | XP_016860022.1 | 3 | 712 | |||
| XM_017004534.1 | XP_016860023.1 | 3 | 712 | |||
| XM_017004535.1 | XP_016860024.1 | 3 | 712 | |||
| XM_017004536.1 | XP_016860025.1 | 3 | 712 | |||
| XM_017004537.1 | XP_016860026.1 | 3 | 712 | |||
| XM_017004538.1 | XP_016860027.1 | 3 | 744 | |||
| XM_017004539.1 | XP_016860028.1 | 3 | 604 | |||
| XM_017004540.1 | XP_016860029.1 | 3 | 744 | |||
| GenBank mRNA | AB037808.1 | - | 3 | 575 | ||
| AK056036.1 | - | 3 | 609 | |||
| AY825268.1 | - | 3 | 272 | |||
| AY825269.1 | - | 3 | 272 | |||
| BC006215.2 | - | 3 | 583 | 3140319 | ||
| BC060855.1 | - | 3 | 601 | 5259789 | ||

PPP4R3B
Genomic Map