siRNA Details
| Gene Symbol: KAT14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020536.4 | NP_065397.3 | 5 | 1182 | ||
| NR_028402.2 | 4 | 620 | ||||
| GenBank mRNA | AK023759.1 | - | 4 | 620 | ||
| AK297597.1 | - | 3 | 276 | |||
| AU140233.1 | - | 4 | 620 | |||
| BC007537.1 | - | 5 | 1086 | 3350378 | ||
| BC009174.2 | - | 4 | 595 | 3844322 | ||

KAT14
Genomic Map