siRNA Details
| Gene Symbol: KAT14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020536.4 | NP_065397.3 | 10 | 2742 | ||
| NR_028402.2 | 9 | 2180 | ||||
| GenBank mRNA | AF252257.1 | - | 5 | 671 | ||
| AK023634.1 | - | 1 | 1504 | |||
| AK023759.1 | - | 9 | 2180 | |||
| AK297597.1 | - | 8 | 1836 | |||
| BC007537.1 | - | 10 | 2646 | 3350378 | ||
| BC009174.2 | - | 9 | 2155 | 3844322 | ||

KAT14
Genomic Map