siRNA Details
| Gene Symbol: ZNF304 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290318.1 | NP_001277247.1 | 2 | 536 | ||
| NM_001290319.1 | NP_001277248.1 | 3 | 617 | |||
| NM_020657.3 | NP_065708.2 | 2 | 536 | |||
| XM_011527145.2 | XP_011525447.1 | 3 | 638 | |||
| XM_017027024.1 | XP_016882513.1 | 2 | 555 | |||
| GenBank mRNA | AJ276316.1 | - | 3 | 431 | ||
| BC111531.1 | - | 3 | 565 | 40012297 | ||
| BC111532.1 | - | 2 | 484 | 40012300 | ||
| DA788435.1 | - | 2 | 534 | |||

ZNF304
Genomic Map