siRNA Details
| Gene Symbol: CLK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020666.2 | NP_065717.1 | 9 | 1081 | ||
| GenBank mRNA | AF294429.1 | - | 8 | 973 | ||
| AK311485.1 | - | 8 | 992 | |||
| BC015942.1 | - | 9 | 1081 | |||
| BC151233.1 | - | 9 | 2295 | |||
| BX647323.1 | - | 5 | 938 | |||
| CR749504.1 | - | 11 | 1960 | |||

CLK4
Genomic Map