siRNA Details
| Gene Symbol: CLK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020666.2 | NP_065717.1 | 10, 11 | 1242 | ||
| GenBank mRNA | AF294429.1 | - | 9, 10 | 1134 | ||
| BC015942.1 | - | 10, 11 | 1242 | |||
| BC136261.1 | - | 10, 11 | 1190 | 9020248 | ||
| BC151233.1 | - | 10, 11 | 2456 | |||
| CR749504.1 | - | 12, 13 | 2121 | |||

CLK4
Genomic Map