siRNA Details
| Gene Symbol: RAB22A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020673.2 | NP_065724.1 | 4 | 501 | ||
| GenBank mRNA | AF091034.2 | - | 4 | 479 | ||
| AJ276210.1 | - | 4 | 466 | |||
| AK091180.1 | - | 4 | 255 | |||
| BC015710.1 | - | 4 | 426 | 3907891 | ||
| BC063457.1 | - | 4 | 485 | 5182786 | ||
| BI457862.1 | - | 4 | 506 | |||
| BT007046.1 | - | 4 | 239 | |||

RAB22A
Genomic Map