siRNA Details
| Gene Symbol: RAB22A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020673.2 | NP_065724.1 | 5 | 556 | ||
| GenBank mRNA | AF091034.2 | - | 5 | 534 | ||
| AJ276210.1 | - | 5 | 521 | |||
| AK091180.1 | - | 5 | 310 | |||
| BC015710.1 | - | 5 | 481 | 3907891 | ||
| BC063457.1 | - | 5 | 540 | 5182786 | ||
| BT007046.1 | - | 5 | 294 | |||

RAB22A
Genomic Map