siRNA Details
| Gene Symbol: TMCC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301036.1 | NP_001287965.1 | 2, 3 | 1136 | ||
| NM_020698.3 | NP_065749.2 | 2, 3 | 1117 | |||
| GenBank mRNA | AB032971.1 | - | 2, 3 | 1046 | ||
| BC040535.1 | - | 2, 3 | 1103 | 5264859 | ||
| BE158711.1 | - | 1, 2 | 83 | |||

TMCC3
Genomic Map