siRNA Details
| Gene Symbol: SLC12A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134771.1 | NP_001128243.1 | 19 | 2573 | ||
| NM_020708.4 | NP_065759.1 | 19 | 2591 | |||
| XM_017027981.1 | XP_016883470.1 | 19 | 2573 | |||
| GenBank mRNA | AB033002.1 | - | 19 | 2406 | ||
| AF208159.1 | - | 19 | 2428 | |||
| AK289758.1 | - | 19 | 2550 | |||
| AK294059.1 | - | 15 | 1768 | |||
| AK295900.1 | - | 13 | 1787 | |||
| BC132668.1 | - | 19 | 2516 | 40146690 | ||
| BC132670.1 | - | 19 | 2516 | 40146692 | ||

SLC12A5
Genomic Map