siRNA Details
| Gene Symbol: PLEKHH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020715.2 | NP_065766.1 | 5, 6 | 554 | ||
| XM_011537009.1 | XP_011535311.1 | 5, 6 | 646 | |||
| XM_017021499.1 | XP_016876988.1 | 4, 5 | 627 | |||
| XM_017021500.1 | XP_016876989.1 | 5, 6 | 646 | |||
| XM_017021501.1 | XP_016876990.1 | 4, 5 | 627 | |||
| XM_017021504.1 | XP_016876993.1 | 1, 2 | 88 | |||
| GenBank mRNA | AB033026.1 | - | 5, 6 | 530 | ||
| AK307428.1 | - | 5, 6 | 646 | |||
| BC146787.1 | - | 5, 6 | 530 | 9007246 | ||

PLEKHH1
Genomic Map