siRNA Details
| Gene Symbol: USP31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020718.3 | NP_065769.3 | 16 | 2717 | ||
| XM_017023496.1 | XP_016878985.1 | 15 | 2628 | |||
| XM_017023497.1 | XP_016878986.1 | 16 | 4243 | |||
| XM_017023498.1 | XP_016878987.1 | 15 | 2030 | |||
| XM_017023499.1 | XP_016878988.1 | 13 | 1834 | |||
| GenBank mRNA | AB033029.1 | - | 5 | 842 | ||
| AJ586135.2 | - | 16 | 2717 | |||
| AK092790.1 | - | 2 | 733 | |||
| AK126447.1 | - | 3 | 1512 | |||
| AK126752.1 | - | 16 | 2401 | |||
| BX648357.1 | - | 7 | 1055 | |||

USP31
Genomic Map