siRNA Details
| Gene Symbol: USP31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020718.3 | NP_065769.3 | 2 | 725 | ||
| XM_017023496.1 | XP_016878985.1 | 2 | 756 | |||
| XM_017023497.1 | XP_016878986.1 | 2 | 2251 | |||
| XM_017023500.1 | XP_016878989.1 | 2 | 756 | |||
| GenBank mRNA | AJ586135.2 | - | 2 | 725 | ||
| AK126752.1 | - | 2 | 409 | |||

USP31
Genomic Map