siRNA Details
| Gene Symbol: INTS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020748.2 | NP_065799.1 | 8 | 1096 | ||
| NR_026641.1 | 8 | 1377 | ||||
| XM_006721987.3 | XP_006722050.1 | 8 | 1103 | |||
| XM_011525058.2 | XP_011523360.1 | 8 | 1383 | |||
| GenBank mRNA | AB033113.1 | - | 8 | 1324 | ||
| AK299032.1 | - | 8 | 1352 | |||
| AL136800.1 | - | 8 | 1096 | |||
| CR533582.1 | - | 7 | 1020 | |||

INTS2
Genomic Map